missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein E/ApoE Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31123G
This item is not returnable.
View return policy
Description
Apolipoprotein E/ApoE Polyclonal specifically detects Apolipoprotein E/ApoE in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Apolipoprotein E/ApoE | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571 | |
| Rabbit | |
| Affinity Purified | |
| Alzheimers Research, Apoptosis, Cancer, Cholesterol Metabolism, Cognition and Behavior, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 348 | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| APOE | |
| Apolipoprotein E/ApoE Antibody made to carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Apolipoprotein E. The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction