missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apolipoprotein D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA514322
This item is not returnable.
View return policy
Description
This antibody is predicted to react with rat and rabbit based on sequence homology.
Apolipoprotein D (APOD) is a component of high density lipoprotein that has no marked similarity to other apolipoprotein sequences. It has a high degree of homology to plasma retinol-binding protein and other members of the alpha 2 microglobulin protein superfamily of carrier proteins, also known as lipocalins. This glycoprotein is closely associated with the enzyme lecithin:cholesterol acyltransferase - an enzyme involved in lipoprotein metabolism.
Specifications
| Apolipoprotein D | |
| Polyclonal | |
| Unconjugated | |
| APOD | |
| AOPDGN; APOD; Apo-D; APO-D protein; Apolipoprotein D | |
| Rabbit | |
| Ammonium Sulfate Precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 11815, 347 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P05090, P51910 | |
| APOD | |
| KLH conjugated synthetic peptide between 26-53 amino acids from the N-terminal region of human APOD. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction