missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein D Mouse anti-Human, Clone: D544, Novus Biologicals™

Product Code. 18316522
Change view
Click to view available options
Quantity:
1000 μg
250 μg
Unit Size:
1000µg
250µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18316522 250 μg 250µg
18342802 1000 μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18316522 Supplier Bio-Techne Supplier No. NBP318285250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Apolipoprotein D Monoclonal antibody specifically detects Apolipoprotein D in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein D
Applications ELISA
Classification Monoclonal
Clone D544
Concentration 0.5 mg/ml
Conjugate Unconjugated
Formulation 0.2 um filtered PBS
Gene Alias apo-D, ApoD, apolipoprotein D
Host Species Mouse
Immunogen Recombinant human Apolipoprotein D
Purification Method Protein G purified
Quantity 250 μg
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 347
Target Species Human
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.