missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein D Mouse anti-Human, Clone: APOD/3414, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07383-100ug
This item is not returnable.
View return policy
Description
Apolipoprotein D Monoclonal specifically detects Apolipoprotein D in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Apolipoprotein D | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| apo-D, ApoD, apolipoprotein D | |
| Human recombinant Apolipoprotein D protein fragment (around aa 21-119) (exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Peptide Array | |
| APOD/3414 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 347 | |
| Store at 4°C. | |
| IgG2c κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction