missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein D Mouse anti-Human, Clone: APOD/3413, Novus Biologicals™

Product Code. 18343707 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343707 20 μg 20µg
18352556 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18343707 Supplier Novus Biologicals Supplier No. NBP31394620UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Apolipoprotein D Monoclonal antibody specifically detects Apolipoprotein D in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein D
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone APOD/3413
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P05090
Gene Alias apo-D, ApoD, apolipoprotein D
Host Species Mouse
Immunogen Human recombinant Apolipoprotein D protein fragment (around aa21-119) (exact sequence is proprietary) (Uniprot: P05090)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 347
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.