missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein C4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-46869-25ul
This item is not returnable.
View return policy
Description
Apolipoprotein C4 Polyclonal antibody specifically detects Apolipoprotein C4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Apolipoprotein C4 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| P55056 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 346 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| APOC4, apo-CIV, ApoC-IV, Apolipoprotein C4, apolipoprotein C-IV | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: QTYYDDHLRDLGPLTKAWFLESKDSLLKKTHSLCPRLVCGDKDQG | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction