missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein B/ApoB Mouse anti-Human, Clone: APOB/4333, Novus Biologicals™
Mouse Monoclonal Antibody
£405.00
Specifications
| Antigen | Apolipoprotein B/ApoB |
|---|---|
| Clone | APOB/4333 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | ELISA, Peptide Array |
Description
Apolipoprotein B/ApoB Monoclonal specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| Apolipoprotein B/ApoB | |
| 0.2 mg/mL | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 338 | |
| IgG1 κ | |
| Protein A or G purified |
| APOB/4333 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100 | |
| Human recombinant Apolipoprotein B/ApoB protein fragment (around aa592-689) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title