missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Apolipoprotein A5 Antibody (1G5G9), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-55454PE
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Apolipoprotein A5 Monoclonal specifically detects Apolipoprotein A5 in Human samples. It is validated for Flow Cytometry.
Tekniske data
| Apolipoprotein A5 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| APOA5 | |
| Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProt# Q6Q788] | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1G5G9 | |
| Flow Cytometry | |
| Apo-AV, ApoA-V, APOAV, Apolipoprotein A5, apolipoprotein A-V, FLJ97995, MGC126836, RAP3MGC126838, Regeneration-associated protein 3 | |
| Mouse | |
| Protein G purified | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 116519 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion