missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein A-IV/ApoA4 Antibody (CL0465), Novus Biologicals™
Mouse Monoclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | Apolipoprotein A-IV/ApoA4 |
|---|---|
| Clone | CL0465 |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695458
|
Novus Biologicals
NBP2-52930-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676517
|
Novus Biologicals
NBP2-52930 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apolipoprotein A-IV/ApoA4 Monoclonal antibody specifically detects Apolipoprotein A-IV/ApoA4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Apolipoprotein A-IV/ApoA4 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| apo-AIV, ApoA-IV, Apolipoprotein A4, apolipoprotein A-IV, MGC142154, MGC142156 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LEGLTFQMKKNAEELKARISASAEELRQRLAPLAEDVRGNLRGNTEGLQKSLAELGGHLDQQVEEFRRRVEPYGENFNKALVQQMEQLRQKLGPHAGDVEGHLSFLEKDLRDKVNSFFSTFKEKESQDKTLSLP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL0465 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| PBS (pH 7.2), 40% Glycerol | |
| 337 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title