missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein A-II/ApoA2 Rabbit anti-Human, Mouse, Rat, Clone: 5F7U1, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Apolipoprotein A-II/ApoA2 Monoclonal antibody specifically detects Apolipoprotein A-II/ApoA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
| Antigen | Apolipoprotein A-II/ApoA2 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 5F7U1 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | APOA2, apoAII, Apo-AII, ApoA-II, Apolipoprotein A2, apolipoprotein A-II |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human Apolipoprotein A-II/ApoA2 (P02652). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?