missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein A-I/ApoA1 Antibody, DyLight 680, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP1-77008FR
This item is not returnable.
View return policy
Description
Apolipoprotein A-I/ApoA1 Polyclonal antibody specifically detects Apolipoprotein A-I/ApoA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| Apolipoprotein A-I/ApoA1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| DyLight 680 | |
| APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, MGC117399 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human ApoA1. The immunogen is located within the first 50 amino acids of ApoA1. Amino Acid Squence: FWQQDEPPQSPWDRRVKD | |
| 0.1 mL | |
| Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
| 335 | |
| Store at 4°C in the dark. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction