missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein A-I/ApoA1 Antibody (2083A) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB36641JF635
This item is not returnable.
View return policy
Description
Apolipoprotein A-I/ApoA1 Monoclonal antibody specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry
Specifications
| Apolipoprotein A-I/ApoA1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, MGC117399 | |
| E. coli-derived recombinant human ApoA1, Asp25-Gln267, Accession # P02647 | |
| 0.1 mL | |
| Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
| 335 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 2083A | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction