missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOL6 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-51929
This item is not returnable.
View return policy
Description
APOL6 Polyclonal specifically detects APOL6 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| APOL6 | |
| Polyclonal | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:2000 | |
| NP_085144.1 | |
| APOL6 | |
| Peptide with sequence C-DDIDKTHKKFTKAN corresponding to internal region according to NP_085144.1. | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| apolipoprotein L, 6, apolipoprotein L6, Apolipoprotein L-VI, APOLVI, APOL-VI, DKFZp667M075, FLJ38562, FLJ90164, MGC57495 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 80830 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction