missing translation for 'onlineSavingsMsg'
Learn More

APOL4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen APOL4
Concentration 0.81 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897023
View Documents
Proteintech
10564-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16887023
View Documents
Proteintech
10564-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is a member of the apolipoprotein L family and may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver. Two transcript variants encoding two different isoforms have been found for this gene. Only one of the isoforms appears to be a secreted protein.
Specifications

Specifications

APOL4
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence
Unconjugated
Rabbit
Human, Mouse
Q9BPW4
80832
APOL4 Fusion Protein Ag0849
Primary
-20°C
APOL4
0.81 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
APOL IV, APOL4, Apolipoprotein L IV, apolipoprotein L, 4, Apolipoprotein L4, APOLIV
APOL4
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.