missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOL1, Mouse anti-Human, Clone: CL0171, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15549.100uL
This item is not returnable.
View return policy
Description
Sequence: SNFLSLAGNTYQLTRGIGKDIRALRRARANLQSVPHASASRPRVTEPISAESGEQVERVNEPSILEMSRGVKLTDVAPVSFFLVLDVVYLVYESKHLHEGAKSETAEELKKVAQELEEKLNILNNSpecifications
| APOL1 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| APO-L/APOL/APOL-I | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| CL0171 | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:2500-1:5000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| apolipoprotein L, 1 | |
| APOL1 | |
| Recombinant protein corresponding to human APOL1. | |
| RUO | |
| 8542 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction