missing translation for 'onlineSavingsMsg'
Learn More

APOL1 Mouse anti-Human, Clone: 1G12D11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66124-1-IG-20UL

 View more versions of this product

Product Code. 16857715

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody recognizes the endogenous ApoL1 of 39-45 kDa in blood lysate. This antibody is specific to APOL1.

Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein L gene family encodes six highly homologous proteins designated apoL-I to -VI, which are associated with large high density type lipoproteins (HDL). The human apoL family maps to chromosome 22q12.1- 13.1 within a 127,000-bp region. apoL has been characterized as a pancreas specific, 383-amino acid, 42 kDa protein that contains a 12-amino acid secretory signal peptide. The apoL genes have TATA-less promoters and contain putative sterol regulatory elements, suggesting that transcription of these genes may be coordinated with that of the low density lipoprotein receptor and genes in pathways involving the synthesis of triglycerides and cholesterol. apoL homologs can undergo 10 fold changes in expression during atherosclerotic changes in vascular endothelial cells, which includes the inflammatory reaction of atherosclerotic lesions.
Specifications

Specifications

APOL1
Monoclonal
1.57 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14791
APOL1
APOL1-Specific Fusion Protein Ag2016
20 μL
Primary
Human
Antibody
IgG2a
Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
1G12D11
Unconjugated
APOL1
APO L, APOL, APOL I, APOL1, Apolipoprotein L, Apolipoprotein L I, apolipoprotein L, 1, Apolipoprotein L1
Mouse
Protein A
RUO
8542
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.