missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APOF Polyclonal Antibody

Product Code. 17783624
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17783624 20 μL 20µL
17730374 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17783624 Supplier Invitrogen™ Supplier No. PA5122230

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

APOF Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The product of this gene is one of the minor apolipoproteins found in plasma. This protein forms complexes with lipoproteins and may be involved in transport and/or esterification of cholesterol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOF
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene APOF
Gene Accession No. Q91V80
Gene Alias AI255964; APOF; Apo-F; Apolipoprotein F; APON; Ba1-666; DKFZp781G18150; Leukemia virus-inactivating factor; lipid transfer inhibitor protein; Liver regeneration-related protein LRRG151; LTIP; LVIF; MGC22520; ovarian and testicular apolipoprotein N; Unknown (protein for MGC:137251)
Gene Symbols APOF
Host Species Rabbit
Immunogen Recombinant protein Apolipoprotein F. The antigen corresponds to amino acid range 111-314 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103161
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.