missing translation for 'onlineSavingsMsg'
Learn More

ApoE4 Antibody (4E4), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18431306 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431306 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431306 Supplier Novus Biologicals Supplier No. NBP149529AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ApoE4 Monoclonal antibody specifically detects ApoE4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen ApoE4
Applications Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 4E4
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571
Host Species Mouse
Immunogen Synthetic peptide made to an internal region of human ApoE4 (within residues 100-150) [Uniprot: P02649]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 348
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.