missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ApoE4 Antibody (4E4), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-49529AF594
This item is not returnable.
View return policy
Description
ApoE4 Monoclonal specifically detects ApoE4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ApoE4 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| APOE | |
| Synthetic peptide made to an internal region of human ApoE4 (within residues 100-150) [Uniprot: P02649] | |
| 0.1 mL | |
| Primary | |
| This antibody is specific for ApoE4. It does not cross-react with the ApoE2 or ApoE3 isoforms. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 40000 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571 | |
| Mouse | |
| Protein G purified | |
| Alzheimers Research, Apoptosis, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 348.0 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction