Learn More
APOE Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18254-1-AP-150UL
This item is not returnable.
View return policy
Description
The APOE antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human APOE. This antibody recognizes human, mouse antigen. The APOE antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
| APOE | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 348 | |
| IgG |
| Western Blot, Immunohistochemistry, Flow Cytometry | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |