missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APOE (R136S Mutant) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119519
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
The protein encoded by this gene is a major apoprotein of the chylomicron. It binds to a specific liver and peripheral cell receptor, and is essential for the normal catabolism of triglyceride-rich lipoprotein constituents. This gene maps to chromosome 19 in a cluster with the related apolipoprotein C1 and C2 genes. Mutations in this gene result in familial dysbetalipoproteinemia, or type III hyperlipoproteinemia (HLP III), in which increased plasma cholesterol and triglycerides are the consequence of impaired clearance of chylomicron and VLDL remnants. [provided by RefSeq, Jun 2016].
Specifications
| APOE (R136S Mutant) | |
| Polyclonal | |
| Unconjugated | |
| APOE | |
| AD2; APO-E; ApoE4; apolipoprotein E; LDLCQ5; LPG | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 348 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.15 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P02649 | |
| APOE | |
| Carrier-protein conjugated synthetic peptide surrounding mutant R136S of human Apolipoprotein E. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction