missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOD Mouse anti-Human, Clone: 1C6D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66215-1-IG-150UL
This item is not returnable.
View return policy
Description
Apolipoprotein D (APOD) is a component of high density lipoprotein that has no marked similarity to other apolipoprotein sequences. It has a high degree of homology to plasma retinol-binding protein and other members of the alpha 2 microglobulin protein superfamily of carrier proteins, also known as lipocalins. This glycoprotein is closely associated with the enzyme lecithin:cholesterol acyltransferase - an enzyme involved in lipoprotein metabolism.Specifications
| APOD | |
| Monoclonal | |
| 0.94 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05090 | |
| APOD | |
| APOD Fusion Protein Ag21422 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1C6D10 | |
| Unconjugated | |
| APOD | |
| Apo D, APOD, apolipoprotein D | |
| Mouse | |
| Protein G | |
| RUO | |
| 347 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction