missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APOC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102480
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total APOC2.
Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein C gene family encodes four homologous proteins designated apoC-I to -IV, which specifically modulate the metabolism of triglyceride-rich lipoproteins. The human apoC-I gene maps to chromosome 19q13.2 and is expressed primarily in the liver where it is activated when monocytes differentiate into macrophages. The human apoC-II gene maps to chromosome 19q13.2 and encodes a 79 amino acid single chain protein that is a necessary cofactor for the activation of lipoprotein lipase, the enzyme that hydrolyzes triglycerides in plasma and transfers the fatty acids to tissues.
Specifications
| APOC2 | |
| Polyclonal | |
| Unconjugated | |
| APOC2 | |
| APC 2; APC2; APO C2; Apo CII; APOC 2; ApoC II; Apoc2; APOC2 protein; ApoCII; APO-CII; apoC-II; Apolipoprotein C II; apolipoprotein C2; Apolipoprotein CII; Apolipoprotein C-II; ApolipoproteinCII; MGC75082; ProapoC-II; Proapolipoprotein C-II; RGD1560725 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11813, 292697, 344 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P02655, Q05020 | |
| APOC2 | |
| A synthesized peptide derived from human APOC2(Accession P02655), corresponding to amino acid residues G2-K52. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction