missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APOBEC3G Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518701
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The Apoliprotein B mRNA-editing, enzyme-catalytic, polypeptide-like (APOBEC) 3 is a multi-isoform member of the cytidine deaminase family of enzymes that act on monomeric nucleoside and nucleotide substrates. Similar to TRIM5-a which targets incoming retroviral capsids, APOBEC3 plays a major role in cellular defense against retroviral infection as at least two isoforms, APOBEC3G and to a lesser extent APOBEC3F, can be incorporated HIV-1 virions and induce hypermutation in the newly synthesized viral DNA and thus destabilize the viral genome. This innate mechanism of retroviral resistance is counteracted by the HIV-1 Vif protein by inducing the ubiquitization and degradation of APOBEC3G; a single amino acid substitution (D128K) blocks APOBEC3G depletion without affecting its inhibitory activity.
Specifications
| APOBEC3G | |
| Polyclonal | |
| Unconjugated | |
| APOBEC3G | |
| A3G; APOBEC3G; APOBEC-related cytidine deaminase; APOBEC-related protein; APOBEC-related protein 9; apolipoprotein B editing enzyme catalytic polypeptide-like 3G; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3G; apolipoprotein B mRNA editing enzyme catalytic subunit 3G; apolipoprotein B mRNA editing enzyme cytidine deaminase; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3G; apolipoprotein B mRNA-editing enzyme catalytic polypeptide 3G; ARCD; ARP9; ARP-9; ASGPR2; ASGP-R2; bK150C2.7; CEM15; CEM-15; CLEC4H2; Deoxycytidine deaminase; dJ494G10.1; DNA dC->dU editing enzyme; DNA dC->dU-editing enzyme APOBEC-3G; FLJ12740; HBXBP; HL-2; MDS019; OTTHUMP00000028911; phorbolin-like protein MDS019 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 60489 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9HC16 | |
| APOBEC3G | |
| Synthetic peptide sequence (DEHSQDLSGRLR) corresponding to the C-terminus amino acids of APOBEC3G (aa 365-377). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction