missing translation for 'onlineSavingsMsg'
Learn More

APOBEC3G Antibody, DyLight 550, Novus Biologicals™

Product Code. 18401903 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401903 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401903 Supplier Novus Biologicals Supplier No. NBP177206R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

APOBEC3G Polyclonal antibody specifically detects APOBEC3G in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOBEC3G
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias APOBEC-related cytidine deaminase, APOBEC-related protein, APOBEC-related protein 9, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3G, ARCD, ARP-9, bK150C2.7, CEM-15, CEM15ARP9, dJ494G10.1, DNA dC->dU-editing enzyme APOBEC-3G, EC 3.5.4, EC 3.5.4.-, EC 3.5.4.5, FLJ12740DNA dC->dU editing enzyme, MDS019, phorbolin-like protein MDS019
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the amino-terminus of human APOBEC3G. The immunogen is located within the first 50 amino acids of APOBEC3G. Amino Acid Squence: RNTVERMYRDTFSYN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 60489
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.