missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOBEC3G Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77206
This item is not returnable.
View return policy
Description
APOBEC3G Polyclonal specifically detects APOBEC3G in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| APOBEC3G | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| APOBEC-related cytidine deaminase, APOBEC-related protein, APOBEC-related protein 9, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3G, ARCD, ARP-9, bK150C2.7, CEM-15, CEM15ARP9, dJ494G10.1, DNA dC->dU-editing enzyme APOBEC-3G, EC 3.5.4, EC 3.5.4.-, EC 3.5.4.5, FLJ12740DNA dC->dU editing enzyme, MDS019, phorbolin-like protein MDS019 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 60489 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 5 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 1 μg/mL, Immunohistochemistry-Paraffin 1 μg/mL | |
| NP_068594 | |
| APOBEC3G | |
| Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the amino-terminus of human APOBEC3G. The immunogen is located within the first 50 amino acids of APOBEC3G. Amino Acid Squence: RNTVERMYRDTFSYN | |
| 0.1 mg | |
| Primary | |
| APOBEC3G antibody will also detect the APOBEC3F isoform. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction