missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APOBEC3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121220
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, U-251MG, MCF7. The target is usually found in the following locations: Nucleus. Immunogen sequence: EEFAYCWENF VYNEGQQFMP WYKFDENYAF LHRTLKEILR YLMDPDTFTF NFNNDPLVLR RRQTYLCYEV ERLDNGTWVL MDQHMGFLCN EAKNLLCGFY.
This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. A hybrid gene results from the deletion of approximately 29.5 kb of sequence between this gene, APOBEC3B, and the adjacent gene APOBEC3A. The breakpoints of the deletion are within the two genes, so the deletion allele is predicted to have the promoter and coding region of APOBEC3A, but the 3' UTR of APOBEC3B. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| APOBEC3B | |
| Polyclonal | |
| Unconjugated | |
| APOBEC3B | |
| A3B; A3Z2_1; A3Z2-Z3; A3Z3; A3Z3_1; APOBEC1L; Apobec3; Apobec3b; Apobec3f; APOBEC3F1; APOBEC3F2; APOBEC3Z2; APOBEC3Z3; apolipoprotein B editing complex 3; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3B; apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3B; apolipoprotein B mRNA editing enzyme catalytic subunit 3B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3F; apolipoprotein B mRNA-editing complex 3; ARCD3; Arp3; ARP4; BC003314; bK150C 2.2; bK150C2.2; Cem15; CEM-15; cytidine deaminase; DJ742C19.2; DNA dC->dU-editing enzyme APOBEC3; DNA dC->dU-editing enzyme APOBEC-3; DNA dC->dU-editing enzyme APOBEC-3B; FLJ21201; phorbolin 2; phorbolin 3; phorbolin-1-related protein; Phorbolin-2/3; PHRBNL; probable DNA dC->dU-editing enzyme APOBEC3; probable DNA dC->dU-editing enzyme APOBEC-3B; recovery from Friend virus 3; Rfv3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9582 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.88 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9UH17 | |
| APOBEC3B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 151-250 of human APOBEC3B (NP_0048915). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction