missing translation for 'onlineSavingsMsg'
Learn More

APOBEC3B Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18385112 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18385112 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18385112 Supplier Novus Biologicals Supplier No. NBP299174AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

APOBEC3B Polyclonal specifically detects APOBEC3B in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOBEC3B
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias APOBEC1L, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B, ARCD3, ARP4, bK150C2.2, cytidine deaminase, DJ742C19.2, EC 3.5.4, EC 3.5.4.-, FLJ21201, phorbolin 3, Phorbolin-1-related protein, phorbolin-2/3, PHRBNLphorbolin 2, probable DNA dC->dU-editing enzyme APOBEC-3B
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human APOBEC3B.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9582
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.