missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APOBEC3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599584
This item is not returnable.
View return policy
Description
The immunogen sequence for PA5-99584 is VKMDQHRGFLHN. Antibody detects endogenous levels of total APOBEC3A.
PHO1 a member of the cytidine deaminase gene family. The PHO1 gene is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. This gene encodes a protein that lacks the zinc binding activity and may be an expressed pseudogene.
Specifications
| APOBEC3A | |
| Polyclonal | |
| Unconjugated | |
| APOBEC3A | |
| A3A; Apobec3; APOBEC3A; apolipoprotein B editing complex 3; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3A; apolipoprotein B mRNA editing enzyme catalytic subunit 3A; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A; apolipoprotein B mRNA-editing complex 3; Arp3; BC003314; bK150C2.1; CEM15; CEM-15; DNA dC->dU-editing enzyme APOBEC-3; DNA dC->dU-editing enzyme APOBEC-3A; pho1; PHO-1; phorbolin 1; Phorbolin1; phorbolin-1; PHRBN; probable DNA dC->dU-editing enzyme APOBEC-3A; recovery from Friend virus 3; Rfv3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 200315, 80287 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31941, Q99J72 | |
| Apobec3, APOBEC3A | |
| A synthesized peptide derived from human APOBEC3A(Accession P31941), corresponding to amino acid residues I26-L76. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction