missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOA5, Mouse anti-Human, Flag, Clone: 2C12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB0040-M01.100ug
This item is not returnable.
View return policy
Specifications
| APOA5 | |
| Monoclonal | |
| FLAG | |
| In PBS, pH 7.4 | |
| APOA-V/APOAV/MGC126836/MGC126838/RAP3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2C12 | |
| The optimal working dilution should be determined by the end user. | |
| apolipoprotein A-V | |
| APOA5 | |
| Recombinant Flag fusion protein corresponding to amino acids 24-366 of human APOA5. | |
| RUO | |
| 116519 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction