missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APOA4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122234
This item is not returnable.
View return policy
Description
APOA4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is an O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but evidence from several in vitro studies indicate that this apolipoprotein is involved in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.
Specifications
| APOA4 | |
| Polyclonal | |
| Unconjugated | |
| APOA4 | |
| APOA4; Apoa-4; APOAIV; apo-AIV; ApoA-IV; Apolipoprotein A4; apolipoprotein AIV; apolipoprotein A-IV; MGC142154; MGC142156 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 337 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P06727 | |
| APOA4 | |
| Recombinant protein Apolipoprotein A4. The antigen corresponds to amino acid range Glu21-Asn235 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction