missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOA4 Mouse anti-Human, Clone: 3A9A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66252-1-IG-150UL
This item is not returnable.
View return policy
Description
Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is an O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but evidence from several in vitro studies indicate that this apolipoprotein is involved in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.Specifications
| APOA4 | |
| Monoclonal | |
| 0.79 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06727 | |
| APOA4 | |
| APOA4 Fusion Protein Ag12454 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 3A9A8 | |
| Unconjugated | |
| APOA4 | |
| APOA4, apolipoprotein A IV | |
| Mouse | |
| Protein G | |
| RUO | |
| 337 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction