Learn More
Invitrogen™ ApoA1 Monoclonal Antibody (532)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MIA1405
Description
By sandwich ELISA, MIA1405 can be used as a detection antibody with Product # 710263 or # 701239 as a coating antibody, to generate a matched pair. Using these matched pairs, recombinant human Apo A-1, but not recombinant mouse Apo A-1, was detected. MIA1405 can be used to detect Apo A-1 from serum samples. To increase sensitivity of sandwich ELISAs with MIA1405, a biotinylated detection antibody followed by Streptavidin-HRP is recommended. By Western blot, MIA1405 detects recombinant human Apo A-1, but not recombinant mouse Apo A-1. MIA1405 is also recommended for detecting endogenous Apo A-1 by Western blot. MIA1405 was formerly sold as a Seradyn product.
Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.
Specifications
| ApoA1 | |
| Monoclonal | |
| 5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P02647, Q00623 | |
| APOA1 | |
| Purified human plasma Apo A-1. | |
| 1 mg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
| 532 | |
| Unconjugated | |
| APOA1 | |
| 1-Sep; 2-Sep; Alp-1; Apo a; APO A1; apo A-1; APO I; apo(a); APOA1; Apoa-1; ApoAI; Apo-AI; apoA-I; Apolipoprotein; Apolipoprotein A1; apolipoprotein A-1; Apolipoprotein AI; apolipoprotein A-I; apolipoprotein A-I precursor; Apolipoprotein A-I(1-242); Brp-14; Ltw-1; Lvtw-1; MGC102525; MGC117399; MGC124929; preproapolipoprotein A-I; prepro-apolipoprotein-AI; ProapoA-I; Proapolipoprotein A-I; Sep-1; Sep2; Sep-2; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7 | |
| Mouse | |
| Protein A | |
| RUO | |
| 11806, 335 | |
| Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.