Learn More
Invitrogen™ ApoA1 Monoclonal Antibody (311)
Description
By sandwich ELISA, MIA1402 can be used as a detection antibody with Product # 710263 or # 701239 as a coating antibody, to generate a matched pair. Using these matched pairs, recombinant human Apo A-1, but not recombinant mouse Apo A-1, was detected. MIA1402 can be used to detect Apo A-1 from serum samples. To increase sensitivity of sandwich ELISAs with MIA1402, a biotinylated detection antibody followed by Streptavidin-HRP is recommended. By Western blot, MIA1402 detects recombinant human Apo A-1, but not recombinant mouse Apo A-1. MIA1402 may not be as successful for detecting endogenous Apo A-1 by Western blot. MIA1402 was formerly sold as a Seradyn product.
Specifications
Specifications
| Antigen | ApoA1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 311 |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | APOA1 |
| Gene Accession No. | P02647, Q00623 |
| Gene Alias | 1-Sep; 2-Sep; Alp-1; Apo a; APO A1; apo A-1; APO I; apo(a); APOA1; Apoa-1; ApoAI; Apo-AI; apoA-I; Apolipoprotein; Apolipoprotein A1; apolipoprotein A-1; Apolipoprotein AI; apolipoprotein A-I; apolipoprotein A-I precursor; Apolipoprotein A-I(1-242); Brp-14; Ltw-1; Lvtw-1; MGC102525; MGC117399; MGC124929; preproapolipoprotein A-I; prepro-apolipoprotein-AI; ProapoA-I; Proapolipoprotein A-I; Sep-1; Sep2; Sep-2; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.