missing translation for 'onlineSavingsMsg'
Learn More

apo-Ipratropium Bromide (mixture of stereoisomers), TRC

Product Code. 30389846
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389846 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389846 Supplier Toronto Research Chemicals Supplier No. TRCI77906625MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ipratropium Bromide EP Impurity F
CAS 17812-46-3
Molecular Formula C20H28NO2.Br
Quantity 25 mg
Synonym 1αH,5αH-Tropanium, 3α-hydroxy-8-isopropyl-, bromide, atropate (8CI), (3-endo)-8-Methyl-8-(1-methylethyl)-3-[(1-oxo-2-phenyl-2-propen-1-yl)oxy]-8-azoniabicyclo[3.2.1]octane bromide, (1R,3r,5S)-8-Methyl-8-(1-methylethyl)-3-[(2-phenylpropenoyl)oxy]-8-azoniabicyclo[3.2.1]octane bromide, (3-endo)-8-Isopropyl-8-methyl-3-[(2-phenylpropenoyl)oxy]-8-azoniabicyclo[3.2.1]octane bromide
InChI Formula InChI=1S/C20H28NO2.BrH/c1-14(2)21(4)17-10-11-18(21)13-19(12-17)23-20(22)15(3)16-8-6-5-7-9-16;/h5-9,14,17-19H,3,10-13H2,1-2,4H3;1H/q+1;/p-1/t17-,18+,19+,21?;
SMILES [Br-].CC(C)[N+]1(C)[C@@H]2CC[C@H]1CC(C2)OC(=O)C(=C)c3ccccc3
IUPAC Name [(1S,5R)-8-isopropyl-8-methyl-8-azoniabicyclo[3.2.1]octan-3-yl] 2-phenylprop-2-enoate;bromide
Molecular Weight (g/mol) 394.34
Formula Weight 393.1303
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.