missing translation for 'onlineSavingsMsg'
Learn More

β-Apo-13-carotenone, MedChemExpress

Product Code. 16494736
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494736 100 mg 100mg
16480047 50 mg 50mg
16443658 10 mg 10mg
16406397 5 mg 5mg
16400959 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494736 Supplier Medchem Express Supplier No. HY101953
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress β-Apo-13-carotenone (D'Orenone) is a naturally occurring β-apocarotenoid functioned as an antagonist of RXRα.

β-Apo-13-carotenone (D'Orenone) is a naturally occurring β-apocarotenoid functioned as an antagonist of RXRα.

Specifications

Chemical Name or Material β-Apo-13-carotenone
Physical Form Oil
CAS 17974-57-1
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H26O
Synonym D'Orenone
Solubility Information DMSO : 110 mg/mL (425.70 mM; Need ultrasonic)
SMILES CC(/C=C/C=C(C)/C=C/C1=C(C)CCCC1(C)C)=O
Molecular Weight (g/mol) 258.4
Formula Weight 258.4
Percent Purity 96.5%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.