missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APM2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£186.00 - £381.00
Specifications
| Antigen | APM2 |
|---|---|
| Dilution | Western Blot 1:400, Immunohistochemistry 10 - 15 ug/ml |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18087435
|
Novus Biologicals
NBP3-11922-25UG |
25 μg |
£186.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18330453
|
Bio-Techne
NBP3-11922-100UG |
100 μg |
£381.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APM2 Polyclonal antibody specifically detects APM2 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| APM2 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| adipose specific 2, apM-2, APM2, chromosome 10 open reading frame 116 | |
| Synthetic peptide derived from human APM2 protein (Uniprot: Q15847) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 1:400, Immunohistochemistry 10 - 15 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS | |
| 10974 | |
| IgG | |
| Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title