missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APM2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Description
APM2 Polyclonal antibody specifically detects APM2 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | APM2 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:400, Immunohistochemistry 10 - 15 ug/ml |
| Formulation | PBS |
| Gene Alias | adipose specific 2, apM-2, APM2, chromosome 10 open reading frame 116 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from human APM2 protein (Uniprot: Q15847) |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?