missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APLP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558732
This item is not returnable.
View return policy
Description
Immunogen sequence: PYVAQEIQEE IDELLQEQRA DMDQFTASIS ETPVDVRVSS EESEEIPPFH PFHPFPALPE NEDTQPELYH PMK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.
APLP2 May play a role in the regulation of hemostasis. The soluble form may have inhibitory properties towards coagulation factors. May interact with cellular G-protein signaling pathways. May bind to the DNA 5'-GTCACATG-3' (CDEI box).
Specifications
| APLP2 | |
| Polyclonal | |
| Unconjugated | |
| APLP2 | |
| AI790698; amyloid beta (A4) precursor-like protein 2; amyloid beta (A4) precursor-like protein 2-like; amyloid beta precursor like protein 2; amyloid precursor protein homolog HSD-2; Amyloid protein homolog; Amyloid protein precursor-like protein 2; amyloid-like protein 2; APLP2; APLP-2; APPH; APPL2; CDEBP; CDEI box-binding protein; sperm membrane protein (YWK-II); sperm membrane protein YWK-II; testicular tissue protein Li 23; WALPLP2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 334 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q06481 | |
| APLP2 | |
| Recombinant protein corresponding to Human APLP2. Recombinant protein control fragment (Product #RP-98353). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction