Learn More
Invitrogen™ APLP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117016
Description
Positive Samples: Mouse brain, Mouse heart, Mouse testis, Rat brain, Rat heart Immunogen sequence: PTNDVDVYFE TSADDNEHAR FQKAKEQLEI RHRNRMDRVK KEWEEAELQA KNLPKAERQT LIQHFQAMVK ALEKEAASEK QQLVETHLAR VEAMLNDRRR MALENYLAAL QSDPPRPHRI LQALRRYVRA ENKDRLHTIR HYQHVLAVDP EKAAQMKSQV MTHLHVIEER RNQSLSLLYK VPYVAQEIQE EIDELLQEQR ADMDQFTASI S.
APLP2 May play a role in the regulation of hemostasis. The soluble form may have inhibitory properties towards coagulation factors. May interact with cellular G-protein signaling pathways. May bind to the DNA 5'-GTCACATG-3' (CDEI box).
Specifications
| APLP2 | |
| Polyclonal | |
| Unconjugated | |
| APLP2 | |
| AI790698; amyloid beta (A4) precursor-like protein 2; amyloid beta (A4) precursor-like protein 2-like; amyloid beta precursor like protein 2; amyloid precursor protein homolog HSD-2; Amyloid protein homolog; Amyloid protein precursor-like protein 2; amyloid-like protein 2; APLP2; APLP-2; APPH; APPL2; CDEBP; CDEI box-binding protein; sperm membrane protein (YWK-II); sperm membrane protein YWK-II; testicular tissue protein Li 23; WALPLP2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11804, 334, 64312 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15943, Q06335, Q06481 | |
| APLP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 370-580 of human APLP2 (NP_0016331). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.