missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APLP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578796
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human SH-SY5Y whole cell, human HepG2 whole cell, human Hela whole cell, rat brain tissue, mouse brain tissue.
APLP2 May play a role in the regulation of hemostasis. The soluble form may have inhibitory properties towards coagulation factors. May interact with cellular G-protein signaling pathways. May bind to the DNA 5'-GTCACATG-3' (CDEI box).
Specifications
| APLP2 | |
| Polyclonal | |
| Unconjugated | |
| APLP2 | |
| AI790698; amyloid beta (A4) precursor-like protein 2; amyloid beta (A4) precursor-like protein 2-like; amyloid beta precursor like protein 2; amyloid precursor protein homolog HSD-2; Amyloid protein homolog; Amyloid protein precursor-like protein 2; amyloid-like protein 2; APLP2; APLP-2; APPH; APPL2; CDEBP; CDEI box-binding protein; sperm membrane protein (YWK-II); sperm membrane protein YWK-II; testicular tissue protein Li 23; WALPLP2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11804, 334, 64312 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P15943, Q06335, Q06481 | |
| APLP2 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human APLP2 (639-656aa EEKVINSKNKVDENMVID). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction