missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APLP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12305-2-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the highly conserved amyloid precursor protein gene family. The encoded protein is a membrane-associated glycoprotein that is cleaved by secretases in a manner similar to amyloid beta A4 precursor protein cleavage. This cleavage liberates an intracellular cytoplasmic fragment that may act as a transcriptional activator. The encoded protein may also play a role in synaptic maturation during cortical development. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| APLP1 | |
| Polyclonal | |
| Unconjugated | |
| APLP1 | |
| Amyloid like protein 1, APLP, APLP 1, APLP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11803, 333 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51693, Q03157 | |
| APLP1 | |
| APLP1 Fusion Protein Ag2951 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction