missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APLP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578795
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Testis Tissue, SGC whole cell, 22RV1 whole cell, MCF-7 whole cell. IHC: Mouse Brain tissue, Rat Brain tissue. Flow: SiHa cell.
This gene encodes a member of the highly conserved amyloid precursor protein gene family. The encoded protein is a membrane-associated glycoprotein that is cleaved by secretases in a manner similar to amyloid beta A4 precursor protein cleavage. This cleavage liberates an intracellular cytoplasmic fragment that may act as a transcriptional activator. The encoded protein may also play a role in synaptic maturation during cortical development. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| APLP1 | |
| Polyclonal | |
| Unconjugated | |
| APLP1 | |
| amyloid beta (A4) precursor-like protein 1; amyloid beta precursor like protein 1; amyloid precursor-like protein 1; amyloid-like protein 1; APLP; APLP 1; APLP1; APLP-1; Aplp1_retired; APLPAPLP-1; C30; RGD1561211 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11803, 333, 502317 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P51693, Q03157 | |
| APLP1 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human APLP1 (82-112aa RRCLRDPQRVLEYCRQMYPELQIARVEQATQ). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction