missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APLP-2 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98659AF594
This item is not returnable.
View return policy
Description
APLP-2 Polyclonal antibody specifically detects APLP-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| APLP-2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP-2, APPHCDEI box-binding protein, APPL2, CDEBP | |
| Produced in rabbits immunized with E. coli-derived Human APLP-2 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 334 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction