missing translation for 'onlineSavingsMsg'
Learn More

APLP-2 Antibody (4B5), Novus Biologicals™

Product Code. 18363958 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363958 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363958 Supplier Novus Biologicals Supplier No. H00000334M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

APLP-2 Monoclonal antibody specifically detects APLP-2 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen APLP-2
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4B5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00373
Gene Alias amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP-2, APPHCDEI box-binding protein, APPL2, CDEBP
Host Species Mouse
Immunogen APLP2 (AAH00373, 41 a.a. ~ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AGTGFAVAEPQIAMFCGKLNMHVNIQTGKWEPDPTGTKSCFETKEEVLQYCQEMYPELQITNVMEANQRVSIDNWCRRDKKQCKSRFVTPFKCLVGEFVSDVLLVPEKCQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 334
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.