missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APLF Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | APLF |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18314456
|
Novus Biologicals
NBP3-15554-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18380544
|
Novus Biologicals
NBP3-15554-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APLF Polyclonal antibody specifically detects APLF in Rat samples. It is validated for Western BlotSpecifications
| APLF | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| APFL, aprataxin- and PNK-like factor, aprataxin and PNK-like factor, aprataxin and PNKP like factor, Apurinic-apyrimidinic endonuclease APLF, C2orf13FLJ16593, chromosome 2 open reading frame 13, EC 4.2.99.18, MGC47799, PALF, PNK and APTX-like FHA domain-containing protein, PNK and APTX-like FHA protein, Xip1, XRCC1-interacting protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-380 of human APLF (NP_775816.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Direct Reversal of DNA Damage | |
| PBS, 50% glycerol, pH7.3 | |
| 200558 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title