missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apixaban open ring amide, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | Des(2-oxo-1-piperidinyl) 6-(4-(5-Amino-5-oxopentyl)) Apixaban |
| CAS | 2187409-01-2 |
| Molecular Formula | C25 H28 N6 O4 |
| Quantity | 2.5 mg |
| Synonym | Des(2-oxo-1-piperidinyl) 6-(4-(5-Amino-5-oxopentyl)) Apixaban, 1H-Pyrazolo[3,4-c]pyridine-3-carboxamide, 6-[4-[(5-amino-5-oxopentyl)amino]phenyl]-4,5,6,7-tetrahydro-1-(4-methoxyphenyl)-7-oxo- (ACI), 6-[4-[(5-Amino-5-oxopentyl)amino]phenyl]-4,5,6,7-tetrahydro-1-(4-methoxyphenyl)-7-oxo-1H-pyrazolo[3,4-c]pyridine-3-carb |
| InChI Formula | InChI=1S/C25H28N6O4/c1-35-19-11-9-18(10-12-19)31-23-20(22(29-31)24(27)33)13-15-30(25(23)34)17-7-5-16(6-8-17)28-14-3-2-4-21(26)32/h5-12,28H,2-4,13-15H2,1H3,(H2,26,32)(H2,27,33) |
| SMILES | COc1ccc(cc1)n2nc(C(=O)N)c3CCN(C(=O)c23)c4ccc(NCCCCC(=O)N)cc4 |
| IUPAC Name | 6-[4-[(5-amino-5-oxopentyl)amino]phenyl]-1-(4-methoxyphenyl)-7-oxo-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxamide |
| Molecular Weight (g/mol) | 476.528 |
| Formula Weight | 476.217 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction