missing translation for 'onlineSavingsMsg'
Learn More

Apixaban, MedChemExpress

Product Code. 16491929
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491929 1 g 1g
16470538 5 mg 5mg
16451916 500 mg 500mg
16437846 100 mg 100mg
16431316 200 mg 200mg
16418078 10 mg 10mg
16403997 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16491929 Supplier Medchem Express Supplier No. HY50667
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Apixaban (BMS-562247-01) is a highly selective, reversible and orally active inhibitor of Factor Xa with Ki of 0.08 nM and 0.17 nM in human and rabbit, respectively. Apixaban is in development for the prevention and treatment of various thromboembolic diseases.

Apixaban (BMS-562247-01) is a highly selective, reversible and orally active inhibitor of Factor Xa with Ki of 0.08 nM and 0.17 nM in human and rabbit, respectively. Apixaban is in development for the prevention and treatment of various thromboembolic diseases.

Specifications

Chemical Name or Material Apixaban
CAS 503612-47-3
Color Pale Yellow
Purity Grade Notes Research
Molecular Formula C25H25N5O4
Quantity 1 g
Synonym BMS-562247-01
Solubility Information DMSO : 50 mg/mL (108.81 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES NC(C1=NN(C2=C1CCN(C2=O)C3=CC=C(C=C3)N4CCCCC4=O)C5=CC=C(C=C5)OC)=O
Molecular Weight (g/mol) 459.5
Formula Weight 459.5
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.