missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APITD1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16630805 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16630805 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16630805 Supplier Invitrogen™ Supplier No. PA5104352

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CENPS.

APITD1 was identified in the neuroblastoma tumour suppressor candidate region on chromosome 1p36. It contains a TFIID-31 domain, similar to that found in TATA box-binding protein-associated factor, TAF(II)31, which is required for p53-mediated transcription activation. This gene was expressed at very low levels in neuroblastoma tumours, and was shown to reduce cell growth in neuroblastoma cells, suggesting that it may have a role in a cell death pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APITD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CENPS
Gene Accession No. Q8N2Z9, Q9D084
Gene Alias 2610040C18Rik; 2810407L01Rik; APITD1; APITD1-CORT; APITD1-CORT protein; APITD1-CORT readthrough; apoptosis-inducing TAF9-like domain-containing protein 1; Apoptosis-inducing TAF9-like domain-containing protein 1 homolog; apoptosis-inducing, TAF9-like domain 1; CENPS; CENP-S; Centromere protein S; FAAP16; FANCM-associated histone fold protein 1; FANCM-interacting histone fold protein 1; Fanconi anemia-associated polypeptide of 16 kDa; MHF1; ortholog of human apoptosis-inducing, TAF9-like domain 1 APITD1
Gene Symbols CENPS
Host Species Rabbit
Immunogen A synthesized peptide derived from human CENPS(Accession Q8N2Z9), corresponding to amino acid residues K99-N138.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100534597, 378708, 69928
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.