Learn More
APIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12089-2-AP-150UL
 View more versions of this product
Product Code. 16869593
  • £349.00 / 150µL
Description
The mammalian homologues of the key cell death gene CED-4 in C. elegans has been identified recently from human and mouse and designated Apaf 1 (for apoptosis protease-activating factor 1). Apaf 1 binds to cytochrome c (Apaf 2) and Caspase-9 (Apaf 3), which leads to Caspase-9 activation. Activated Caspase-9 in turn cleaves and activates Caspase-3 that is one of the key proteases, being responsible for the proteolytic cleavage of many key proteins in apoptosis. Recently, Cho et al (4) have identified a new Apaf 1 Interactiong Protein (APIP) also known as CG129 and MMRP19, as a negative regulator of ischemic injury. APIP competes with Caspase-9 binding site of Apaf 1. APIP is predicted to code for a 204 amino acid. An isoform of APIP, APIP2 encodes a 242 amino acid protein, which is an alternative splicing variant differing in its N-terminus from APIP. APIP transcript is ubiquitously expressed in most adult tissue with high expression in skeletal muscle, heart, and kidney.
Specifications
Specifications
APIP
Polyclonal
Unconjugated
APIP
APAF1 interacting protein, APIP, APIP2, CGI 29, CGI29, MMRP19, MTRu 1 P dehydratase
Rabbit
Antigen Affinity Chromatography
RUO
295961, 51074, 56369
-20°C
Liquid
Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96GX9, Q9WVQ5
APIP
APIP Fusion Protein Ag2729
150 μL
Primary
Human, Mouse, Rat